Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT4 Antibody (OTI3D11), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271203AF647
Description
CNOT4 Monoclonal specifically detects CNOT4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
CNOT4 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CNOT4 | |
Recombinant protein fragment corresponding to amino acids 190-455 of human CNOT4 (NP_037448) produced in E.coli. | |
0.1 mL | |
4850.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI3D11 | |
Western Blot, Flow Cytometry | |
CCR4-associated factor 4, CCR4-NOT transcription complex, subunit 4, E3 ubiquitin-protein ligase CNOT4, EC 6.3.2.-, NOT4yeast) homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction