Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT4 Monoclonal Antibody (OTI4B2), TrueMAB™, OriGene
SDP

Catalog No. 502540299
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0299 30 μL
50-167-5160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0299 Supplier OriGene Supplier No. TA504474S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CNOT4 Monoclonal antibody specifically detects CNOT4 in Human samples. It is validated for Western Blot

Specifications

Antigen CNOT4
Applications Western Blot
Classification Monoclonal
Clone OTI4B2
Concentration 2.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CNOT4
Gene Accession No. O95628
Gene Alias CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; CCR4-NOT transcription complex subunit 4 L homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex, subunit 4; CCR4-NOT transcription complex, subunit 4 isoform a; CLONE243; cnot4; cnot4.S; E3 ubiquitin-protein ligase CNOT4; hypothetical protein; negative regulator of transcription 4; Not4; NOT4 (negative regulator of transcription 4, yeast) homolog; Not4h; Not4hp; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp; XELAEV_18019807mg
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 190-455 of human CNOT4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4850
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.