Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT4 Monoclonal Antibody (OTI4C7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504475S


Description
CNOT4 Monoclonal antibody specifically detects CNOT4 in Human samples. It is validated for Western BlotSpecifications
CNOT4 | |
Monoclonal | |
0.56 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95628 | |
Mouse | |
Affinity chromatography | |
RUO | |
4850 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI4C7 | |
Unconjugated | |
CNOT4 | |
CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; CCR4-NOT transcription complex subunit 4 L homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex, subunit 4; CCR4-NOT transcription complex, subunit 4 isoform a; CLONE243; cnot4; cnot4.S; E3 ubiquitin-protein ligase CNOT4; hypothetical protein; negative regulator of transcription 4; Not4; NOT4 (negative regulator of transcription 4, yeast) homolog; Not4h; Not4hp; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp; XELAEV_18019807mg | |
Human recombinant protein fragment corresponding to amino acids 190-455 of human CNOT4 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction