Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNOT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100444
Description
Antibody detects endogenous levels of total CNOT6.
Poly(A) nuclease with 3'-5' RNase activity. Catalytic component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Involved in mRNA decay mediated by the major-protein-coding determinant of instability (mCRD) of the FOS gene in the cytoplasm. In the presence of ZNF335, enhances ligand-dependent transcriptional activity of nuclear hormone receptors, including RARA. The increase of ligand-dependent ESR1-mediated transcription is much smaller, if any. Mediates cell proliferation and cell survival and prevents cellular senescence.
Specifications
CNOT6 | |
Polyclonal | |
Unconjugated | |
CNOT6 | |
carbon catabolite repression 4 protein; carbon catabolite repressor protein 4 homolog; Ccr4; CCR4 carbon catabolite repression 4-like; Ccr4a; CCR4-NOT transcription complex subunit 6; CCR4-NOT transcription complex, subunit 6; Cnot6; Cytoplasmic deadenylase; Kiaa1194; LOW QUALITY PROTEIN: CCR4-NOT transcription complex subunit 6; RGD1310783 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
104625, 287249, 57472 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6AXU9, Q8K3P5, Q9ULM6 | |
CNOT6 | |
A synthesized peptide derived from human CNOT6(Accession Q9ULM6), corresponding to amino acid residues Y495-L545. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction