Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNOT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117878 Supplier Invitrogen™ Supplier No. PA5117878
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNOT7 Polyclonal Antibody for Western Blot, IP

CNOT7 (CCR4-NOT transcription complex subunit 7) has 3'-5' poly(A) exoribonuclease activity for synthetic poly(A) RNA substrate. Its function seems to be partially redundant with that of CNOT8. CNOT7 is a catalytic component of the CCR4-NOT complex, which is one of the major cellular mRNA deadenylases, and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation, and general transcription regulation. During miRNA-mediated repression, the complex seems to act as a transcriptional repressor during translational initiation, as well. Additional complex functions may be a consequence of its influence on mRNA expression. CNOT7 associates with members of the BTG family such as TOB1 and BTG2 and is required for their anti-proliferative activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOT7
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CNOT7
Gene Accession No. Q9UIV1
Gene Alias AU022737; BTG1-binding factor 1; CAF1; CAF-1; Caf1a; carbon catabolite repressor protein (CCR4)-associative factor 1; catabolite repressor protein (CCR4)-associative factor 1; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 7; CCR4-NOT transcription complex subunit-like protein; CCR4-NOT transcription complex, subunit 7; CNOT7; fd59c07; fj07d05; hCAF-1; hypothetical protein LOC508055; Pop2; POP2 homolog; wu:fd59c07; wu:fj07d05; zgc:153168
Gene Symbols CNOT7
Host Species Rabbit
Immunogen E. coli-derived Human CNOT7 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29883
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.