Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNOT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562399 Supplier Invitrogen™ Supplier No. PA562399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CGRLYGLGTG VAQKQNEDVD SAQEKMSILA IINNMQQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOT8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNOT8
Gene Accession No. Q9UFF9
Gene Alias 1500015I04Rik; 1810022F04Rik; AA536816; AU015770; AU043059; CAF1; Caf1b; CAF1-like protein; CAF2; CALIF; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; CNOT8; h CAF1-like protein; hCAF1; PGK promoter directed over production; POP2; wu:fe49a05; zgc:63844
Gene Symbols CNOT8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNOT8. Recombinant protein control fragment (Product #RP-101388).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.