Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989034
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9034 Supplier Bioss Supplier No. BS9388R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.

Specifications

Antigen CNOT8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNOT8
Gene Accession No. Q9UFF9
Gene Alias 1500015I04Rik; 1810022F04Rik; AA536816; AU015770; AU043059; BOS_8375; CAF1; Caf1b; CAF1-like protein; CAF2; CALIF; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; CCR4-NOT transcription complex, subunit 8-like; Cnot8; h CAF1-like protein; hCAF1; LOC100350728; PGK promoter directed over production; POP2; wu:fe49a05; zgc:63844
Gene Symbols CNOT8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CNOT8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9337
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.