Learn More
Invitrogen™ CNP Monoclonal Antibody (347625)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ANP precursor or rhBNP precursor is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CNP |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 347625 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CNP |
| Gene Accession No. | P09543 |
| Gene Alias | 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; CNP1; CNP2; CNP-22; CNP-29; CNP-53; CNPase; C-type natriuretic peptide; C-type natriuretic peptide/factor; HLD20; lbab; natriuretic peptide C; natriuretic peptide precursor C; natriuretic peptide precursor type C; natriuretic peptide type C; NPPC; snake venom metalloprotease inhibitor 02D01-like; SVSP15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.