Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103119
Description
Antibody detects endogenous levels of total CNP-53.
CNP (2',3'-Cyclic Nucleotide 3' Phosphodiesterase) is a Protein Coding gene. Diseases associated with CNP include Leukodystrophy, Hypomyelinating, 20 and Myopia 2, Autosomal Dominant. Among its related pathways are MECP2 and associated Rett syndrome and Glial cell differentiation. Gene Ontology (GO) annotations related to this gene include RNA binding and cyclic nucleotide binding.
Specifications
CNP | |
Polyclonal | |
Unconjugated | |
CNP | |
2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; CNP1; CNP2; CNP-22; CNP-29; CNP-53; CNPase; C-type natriuretic peptide; C-type natriuretic peptide/factor; HLD20; lbab; natriuretic peptide C; natriuretic peptide precursor C; natriuretic peptide precursor type C; natriuretic peptide type C; NPPC; snake venom metalloprotease inhibitor 02D01-like; SVSP15 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1267, 12799, 25275 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P09543, P13233, P16330 | |
CNP | |
A synthesized peptide derived from human NPPC(Accession P23582), corresponding to amino acid residues S81-C126. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction