Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ proCNP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557241 Supplier Invitrogen™ Supplier No. PA557241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKVPRTPPAE ELAEPQAAGG GQKKGDKAPG GGGANLKGDR SRLLRDLRVD TKSRAAWARL LQEHPNARKY KGANKKGLSK GCFGLKLDRI GSMSGLGC Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.

Natriuretic peptides comprise a family of 3 structurally related molecules: atrial natriuretic peptide (ANP; MIM 108780), brain natriuretic peptide (BNP; MIM 600295), and C-type natriuretic peptide, CNP, encoded by a gene symbolized NPPC. These peptides possess potent natriuretic, diuretic, and vasodilating activities and are implicated in body fluid homeostasis and blood pressure control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen proCNP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nppc
Gene Accession No. P23582
Gene Alias Cnp; CNP2; CNP-22; CNP-29; CNP-53; C-type natriuretic peptide; lbab; natriuretic peptide C; natriuretic peptide precursor C; natriuretic peptide precursor type C; natriuretic peptide type C; NPPC
Gene Symbols Nppc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNP. Recombinant protein control fragment (Product #RP-95804).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.