Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNPase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587696 Supplier Invitrogen™ Supplier No. PA587696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKTAWRLDCA QLKEKNQWQL SADDLKKLKP GLEKDFLPLY FGWFLTKKSS ETLRKAGQVF LEELGNHKAF KKELRQFVPG DEPREKMDLV TYFGKRPPGV LHCTTKFCDY GKAPGAEEYA QQDVLKKSYS KAFTLTISAL FVTPKTTGAR VELSEQQLQL WPSDVDKLSP TDNLPRGSRA HITLGCAADV EAVQTGLDLL EILRQEKGGS RGEEVGELSR GKLYSLGNGR WMLTLAKNME VRAIFTGYYG KGKPVPTQGS RKGGALQSCT II Positive Samples: HepG2, 293T, Rat brain, Mouse brain; Cellular Location: Lipid-anchor, Melanosome, Membrane.

CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CNP
Gene Accession No. P09543, P13233, P16330
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1
Gene Symbols CNP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-421 of human CNPase (NP_1491243).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1267, 12799, 25275
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.