Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNPase Monoclonal Antibody (CL2872)
GREENER_CHOICE

Catalog No. PIMA531375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531375 Supplier Invitrogen™ Supplier No. MA531375
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: PGVLHCTTKF CDYGKAPGAE EYAQQDVLKK SYSKAFTLTI SALFVTPKTT GARVELSEQQ LQLWPSDVDK LSPTDNLPRG SRAHITL Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 76%.

CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2872
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNP
Gene Accession No. P09543, P13233, P16330
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1
Gene Symbols CNP
Host Species Mouse
Immunogen Recombinant protein corresponding to Human CNP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1267, 12799, 25275
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.