Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNPase Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA5143564
Description
CNPase Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
Specifications
CNPase | |
Polyclonal | |
Unconjugated | |
CNP | |
2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1 | |
Chicken | |
100 μL | |
Primary | |
Human, Mouse, Rat, Bovine, Pig, Horse | |
Antibody | |
IgY |
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
PBS with 0.02% sodium azide | |
P06623, P09543, P13233, P16330 | |
CNP | |
Full length human recombinant protein. | |
RUO | |
100066401, 100511587, 1267, 12799, 25275, 280752 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction