Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNPase Polyclonal Antibody
Rabbit Polyclonal Antibody
$514.50 - $1763.00
Specifications
Antigen | CNPase |
---|---|
Concentration | Conc. Not Determined |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143503
|
Invitrogen™
PA5143503 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143601
|
Invitrogen™
PA5143601 |
100 μL |
Each for $514.50
|
|
|||||
Description
CNPase Polyclonal Antibody for Western Blot, ICC/IF, IHC
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.Specifications
CNPase | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
CNP | |
2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1 | |
CNP | |
Primary |
Conc. Not Determined | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
Whole serum with 5mM sodium azide | |
P09543, P13233, P16330 | |
1267, 12799, 25275 | |
Full length human recombinant protein expressed in E. coli. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title