Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980624
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0624 Supplier Bioss Supplier No. BS1000R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.

Specifications

Antigen CNPase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNP
Gene Accession No. P09543
Gene Alias 2′, 3′ cyclic nucleotide 3′ phosphohydrolase; 2′,3′-cyclic nucleotide 3′ phosphodiesterase; 2′,3′-cyclic nucleotide 3′ phosphodiesterase; 2′,3′-cyclic-nucleotide 3′-phosphodiesterase; 2′,3′-cyclic-nucleotide 3′-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1
Gene Symbols CNP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CNPase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1267
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.