Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNPPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561926 Supplier Invitrogen™ Supplier No. PA561926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRRLYYGWDW EADCSLEELS SPVADIAVEL LQKAAPSPIR RLQKKYVAHV SREACISPCA MMLALVYIER LRHRNPDYLQ HVSSSDLFLI SMMVASKYLY DEGEEEEVFN DEWGAAGGVA VPTLNALERG FLSAMDWHLY TDPREI Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

CNPPD1 gene ontology annotations related to this gene include regulation of cyclin-dependent protein serine/threonine kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPPD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNPPD1
Gene Accession No. Q9BV87
Gene Alias 1810031K17Rik; AU043960; C2H2orf24; C2orf24; CDABP0125; CGI-57; chromosome 2 open reading frame 24; Cnppd1; cyclin Pas1/PHO80 domain containing 1; cyclin Pas1/PHO80 domain-containing protein 1; hypothetical protein LOC507473; hypothetical protein MGC:72616; LOC27013; protein CNPPD1; RGD735175; Similar to hypothetical protein CGI-57; wu:fa05h07; zgc:55857
Gene Symbols CNPPD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNPPD1. Recombinant protein control fragment (Product #RP-89714).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27013
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.