Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNPY2/MSAP Mouse anti-Human, Mouse, Rat, Clone: 1G8G11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CNPY2/MSAP |
|---|---|
| Clone | 1G8G11 |
| Concentration | 1.13 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6607
![]()
|
Proteintech Group Inc
661731IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to CNPY2.
CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.Specifications
| CNPY2/MSAP | |
| 1.13 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| canopy 2 homolog (zebrafish), CNPY2, CNPY2,MSAP, HP10390, MSAP, Protein canopy homolog 2, TMEM4, Transmembrane protein 4, ZSIG9 | |
| Cnpy2 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1G8G11 | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q9QXT0, Q9Y2B0 | |
| 10330, 56530, 685814 | |
| CNPY2, MSAP Fusion Protein Ag6363 | |
| Primary | |
| -20°C | |
| CNPY2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title