Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNPY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118996
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118996 200 μL
PIPA5122086 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118996 Supplier Invitrogen™ Supplier No. PA5118996
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNPY2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPY2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CNPY2
Gene Accession No. Q9QXT0, Q9Y2B0
Gene Alias 5330432A10Rik; AW229003; canopy 2 homolog; canopy 2 homolog (zebrafish); canopy FGF signaling regulator 2; Cnpy2; D10Bwg1546e; HP10390; MIR-interacting saposin-like protein; MSAP; Protein canopy homolog 2; putative secreted protein Zsig9; Tmem4; transmembrane protein 4; UNQ1943/PRO4426; Zsig9
Gene Symbols CNPY2
Host Species Rabbit
Immunogen Recombinant protein Canopy 2 Homolog. The antigen corresponds to amino acid range 21-182 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100151912, 10330, 56530, 685814
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.