Learn More
CNPY2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305684AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.Specifications
CNPY2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9QXT0, Q9Y2B0 | |
CNPY2 | |
Between 100 and 150 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CNPY2 | |
HP10390, MSAP, TMEM4, ZSIG9, MIR-interacting saposin-like protein, protein canopy homolog 2, putative secreted protein Zsig9, transmembrane protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10330, 56530 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.