Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNPY3 Polyclonal Antibody

CNPY3 Polyclonal Antibody
Supplier: Thermo Scientific PA553443
Description
CNPY3 Polyclonal Antibody for Western Blot, IHC
Specifications
| CNPY3 | |
| Polyclonal | |
| Unconjugated | |
| CNPY3 | |
| 1600025D17Rik; 2410050O22Rik; AI413153; CAG repeat containing; CAG/CTG 5; CAG4A; canopy 3 homolog; canopy 3 homolog (zebrafish); canopy FGF signaling regulator 3; CNPY3; CTG repeat protein 4a; CTG4A; ERDA5; expanded repeat-domain protein CAG/CTG 5; HSPC084; Prat4a; protein associated with TLR4; protein associated with Toll-like receptor 4A; protein canopy homolog 3; Tnrc5; trinucleotide repeat containing 5; trinucleotide repeat-containing gene 5 protein; Unknown (protein for MGC:140466); UNQ1934/PRO4409 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BT09 | |
| CNPY3 | |
| Recombinant protein corresponding to Human CNPY3. Recombinant protein control fragment (Product #RP-91634). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction