Learn More
Invitrogen™ CNPY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596600
Description
Immunogen sequence: GPSQAGAEEN DWVRLPSKCE VCKYVAVELK SAFEETGKTK EVIGTGYGIL DQKASGVKYT KSDLRLIEVT ETICKRLLDY SLHKERTGSN RFAKGMSETF ETLHNLVHKG VKVVMDIPYE LWNETSAEVA DLKKQCDVLV EEFEEVIEDW YRNHQEEDLT EFLCANHVLK GKDTSCLAEQ WSGKKGDTAA LGGKKSKKKS SRAKAAGGRS SSSKQRKELG GLEGDPSPEE DEGIQKASPL THSPPDEL; Positive Samples: HepG2, MCF7, THP-1, HL-60, U-937; Cellular Location: Endoplasmic reticulum.
PRAT4A is a novel protein belonging to the canopy family with a saposin B-type domain. It is an accessory molecule required for microbial recognition by Toll-like receptor (TLR), and mediates TLR dependent immune responses. Though the exact function of PRAT4A is still unknown, reports suggest that it is associated with the immature form of TLR4 and regulates the cell surface expression of this TLR. PRAT4A, a secreted protein, also has a role in TLR4 maturation and glycosylation. PRATA4 is usually present in endoplasmic reticulum and plays an active role in regulating the subcellular distribution and response of multiple TLRs and is required for both innate and adaptive immune responses.
Specifications
CNPY3 | |
Polyclonal | |
Unconjugated | |
CNPY3 | |
1600025D17Rik; 2410050O22Rik; AI413153; CAG repeat containing; CAG/CTG 5; CAG4A; canopy 3 homolog; canopy 3 homolog (zebrafish); canopy FGF signaling regulator 3; CNPY3; CTG repeat protein 4a; CTG4A; ERDA5; expanded repeat-domain protein CAG/CTG 5; HSPC084; Prat4a; protein associated with TLR4; protein associated with Toll-like receptor 4A; protein canopy homolog 3; Tnrc5; trinucleotide repeat containing 5; trinucleotide repeat-containing gene 5 protein; Unknown (protein for MGC:140466); UNQ1934/PRO4409 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10695, 72029 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BT09, Q9DAU1 | |
CNPY3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 31-278 of human CNPY3 (NP_0065772). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.