Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNPY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598919
Description
CNPY3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
PRAT4A is a novel protein belonging to the canopy family with a saposin B-type domain. It is an accessory molecule required for microbial recognition by Toll-like receptor (TLR), and mediates TLR dependent immune responses. Though the exact function of PRAT4A is still unknown, reports suggest that it is associated with the immature form of TLR4 and regulates the cell surface expression of this TLR. PRAT4A, a secreted protein, also has a role in TLR4 maturation and glycosylation. PRATA4 is usually present in endoplasmic reticulum and plays an active role in regulating the subcellular distribution and response of multiple TLRs and is required for both innate and adaptive immune responses.
Specifications
CNPY3 | |
Polyclonal | |
Unconjugated | |
CNPY3 | |
1600025D17Rik; 2410050O22Rik; AI413153; CAG repeat containing; CAG/CTG 5; CAG4A; canopy 3 homolog; canopy 3 homolog (zebrafish); canopy FGF signaling regulator 3; CNPY3; CTG repeat protein 4a; CTG4A; ERDA5; expanded repeat-domain protein CAG/CTG 5; HSPC084; Prat4a; protein associated with TLR4; protein associated with Toll-like receptor 4A; protein canopy homolog 3; Tnrc5; trinucleotide repeat containing 5; trinucleotide repeat-containing gene 5 protein; Unknown (protein for MGC:140466); UNQ1934/PRO4409 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10695 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BT09 | |
CNPY3 | |
Recombinant Human Protein canopy homolog 3 protein (31-278AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction