Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPY4 Polyclonal Antibody, Abbomax

Catalog No. 502204574
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4574 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4574 Supplier Abbomax Inc Supplier No. 630320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Kidney Cellular location: Secreted.

CNPY4 Polyclonal antibody specifically detects CNPY4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CNPY4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene CNPY4
Gene Accession No. Q8BQ47, Q8N129
Gene Alias 2610019P18Rik; canopy 4 homolog; canopy 4 homolog (zebrafish); canopy FGF signaling regulator 4; Cnpy; Cnpy4; Prat4b; PRotein Associated with Tlr4; Protein canopy homolog 4; PSEC0237; RGD1307636; UNQ1909/PRO4354
Host Species Rabbit
Immunogen A synthetic peptides close to C-terminus of human CNPY4 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245812, 363886, 66455
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.