Learn More
Invitrogen™ CNR2 Recombinant Rabbit Monoclonal Antibody (13H43L20)
Description
This antibody is predicted to react with Rat, Bovine.
Specifications
Specifications
| Antigen | CNR2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 13H43L20 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | CNR2 |
| Gene Accession No. | P34972, P47936 |
| Gene Alias | Cannabinoid Receptor 2; cannabinoid receptor 2 (macrophage); CB2; CB-2; CB2 receptor; CB2A; CB2B; CB2C; CB2R; CB2-R; CNR2; CNR2C; CNR3; CX5; HCB2; HGNC:2160; OTTHUMP00000044841; peripheral cannabinoid receptor; rCB2; RP11-4M23.1; testis-dominant CNR2 isoform CB2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.