Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNRIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534546
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534546 Supplier Invitrogen™ Supplier No. PA534546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-34546 can be used with blocking peptide PEP-1589.

The CNRIP1 (cannabinoid receptor interacting protein 1) protein is a G-protein coupled receptor which interacts with the C-terminal tail of cannabinoid receptor 1 (CB1) and is thought to play a role in synaptic plasticity, analgesia, appetite, and neuroprotection. One isoform of CNRIP1, CNRIP1a, modulates the constitutive CB1 receptor activity in the central nervous system (CNS), while the role of the shorter isoform CNRIP1b is yet unknown. CNRIP1 has been suggested as a potential target for CNS drug discovery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNRIP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CNRIP1
Gene Accession No. Q5M7A7, Q5M8N0, Q96F85
Gene Alias 1500041B16Rik; 3110054C06Rik; 5330437A18Rik; AI854501; C2orf32; cannabinoid receptor CB1-interacting protein 1; cannabinoid receptor interacting protein 1; CB1 cannabinoid receptor interacting; CB1 cannabinoid receptor-interacting protein 1; Cnrip1; CRIP1; CRIP-1; Crip1a; RGD1308373
Gene Symbols CNRIP1
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of human CNRIP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25927, 364208, 380686
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.