Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNRIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CNRIP1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1206
![]() ![]() |
Proteintech Group Inc
168271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The CNRIP1 (cannabinoid receptor interacting protein 1) protein is a G-protein coupled receptor which interacts with the C-terminal tail of cannabinoid receptor 1 (CB1) and is thought to play a role in synaptic plasticity, analgesia, appetite, and neuroprotection. One isoform of CNRIP1, CNRIP1a, modulates the constitutive CB1 receptor activity in the central nervous system (CNS), while the role of the shorter isoform CNRIP1b is yet unknown. CNRIP1 has been suggested as a potential target for CNS drug discovery.Specifications
CNRIP1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5M7A7, Q5M8N0, Q96F85 | |
25927, 364208, 380686 | |
CNRIP1 Fusion Protein Ag10428 | |
Primary | |
-20°C | |
CNRIP1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C2orf32, CNRIP1, CRIP 1, CRIP1 | |
CNRIP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title