Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNST Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB181932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181932 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181932 Supplier Novus Biologicals Supplier No. NBP306530B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNST Polyclonal antibody specifically detects CNST in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CNST
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias C1orf71, consortin, connexin sorting protein, FLJ32001, MGC18089, PPP1R64, protein phosphatase 1, regulatory subunit 64
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CNST fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163882
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.