Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552405 Supplier Invitrogen™ Supplier No. PA552405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKEGGATTKE SESKTCLGTE SSKESQHTVE PLGSSPCCHQ MDVQTDSPSL SVTAGKDHME ELLCSAEATL ALHTQSSETA GSPSGPDSSE DACEDDSRLQ LAQTEACQDV ARIEGIAEDP KVFLSSKSKT EPLISPGCDR IPPAL Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 49%.

Required for targeting of connexins to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNST
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNST
Gene Accession No. Q6PJW8
Gene Alias 9630058J23Rik; C1orf71; CNST; Consortin; consortin, connexin sorting protein; PPP1R64; protein phosphatase 1, regulatory subunit 64
Gene Symbols CNST
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNST. Recombinant protein control fragment (Product #RP-89727).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163882
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.