Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNTD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145023
Description
Immunogen sequence: MDGPMRPRSA SLVDFQFGVV ATETIEDALL HLAQQNEQAV REASGRLGRF REPQIVEFVF LLSEQWCLEK SVSYQAV Antigen sequence identity: Mouse 81%; Rat 82%.
CNTD1 is a protein coding gene.
Specifications
CNTD1 | |
Polyclonal | |
Unconjugated | |
CNTD1 | |
1700051C09Rik; 4921513J16Rik; 5430417M23Rik; CNTD; Cntd1; cyclin N-terminal domain containing 1; cyclin N-terminal domain-containing protein 1; RGD1564542 | |
Rabbit | |
Affinity chromatography | |
RUO | |
124817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N815 | |
CNTD1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1-77. Recombinant protein control fragment (Product #RP-110123). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction