Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9306 Supplier Bioss Supplier No. BS9740R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNTD2 gene ontology annotations related to this gene include mitotic cell cycle; positive regulation of cell cycle; positive regulation of cell proliferation; protein phosphorylation; regulation of cyclin-dependent protein serine/threonine kinase activity; regulation of mitotic nuclear division.

Specifications

Antigen CNTD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNTD2
Gene Accession No. Q9H8S5
Gene Alias CCNP; CNTD2; cyclin N-terminal domain containing 2; Cyclin N-terminal domain-containing protein 2; cyclin P
Gene Symbols CNTD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CNTD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79935
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.