Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTF Antibody (121), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290839H
Description
CNTF Monoclonal specifically detects CNTF in Rat samples. It is validated for ELISA.Specifications
CNTF | |
Monoclonal | |
HRP | |
ciliary neurotrophic factor, HCNTF | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CNTF (P20294; Met1-Met200). | |
0.1 mL | |
Biologically Active Proteins, Neuroscience | |
1270 | |
Rat | |
Purified |
ELISA | |
121 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CNTF Mouse LIF Mouse OSM Rat IL6 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction