Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNTF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120667
This item is not returnable.
View return policy
Description
Positive test controls include: Mouse skeletal muscle. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: ARKIRSDLTA LTESYVKHQG LNKNINLDSA DGMPVASTDQ WSELTEAERL Q.
The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.
Specifications
| CNTF | |
| Polyclonal | |
| Unconjugated | |
| CNTF | |
| AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1270, 12803, 25707 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P20294, P26441, P51642 | |
| CNTF | |
| A synthetic peptide corresponding to a sequence within amino acids 24-74 of human CNTF (NP_000605.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction