Learn More
Invitrogen™ CNTFR Polyclonal Antibody

Description
Positive Control: Mouse cerebellum tissue lysate, rat brain tissue lysate, NIH/3T3 cell lysate, human fetal skeletal muscle tissue, mouse heart tissue, Hela. Subcellular Location: Cell membrane.
Specifications
Specifications
| Antigen | CNTFR |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Cntfr |
| Gene Accession No. | O88507, P26992, Q08406 |
| Gene Alias | ciliary neurotrophic factor receptor; ciliary neurotrophic factor receptor alpha; ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; Cntfr; CNTFR alpha; CNTFRALPHA; CNTFR-alpha; MGC1774 |
| Gene Symbols | Cntfr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.