Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547437
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547437 Supplier Invitrogen™ Supplier No. PA547437
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, greater than 50% cross-reactivity with recombinant rat CNTF sR alpha is observed and less than 5% cross-reactivity with recombinant human (rh) IL-2 sR gamma and rhIL-6 sR is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

This gene encodes a hematopoeitin/interferon-class receptor belonging to the cytokine superfamily of receptors. The encoded gene product represents the CNTF-specific alpha subunit of a heterotrimer forming the CNTF receptor complex, which also includes LIFR and gp130. The receptor is attached to the membrane by a glycosyl-phosphatidylinositol linkage and contains an immunoglobulin-like C2-type domain and a fibronectin type-III domain. Signal transduction requires that CNTF bind first to this alpha component, which permits the recruitment of gp130 and LIFR beta to form the tripartite receptor complex. Signal transduction stimulates gene expression, cell survival or differentiation in a variety of neuronal cell types. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTFR
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Cntfr
Gene Accession No. P26992
Gene Alias ciliary neurotrophic factor receptor; ciliary neurotrophic factor receptor alpha; ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; Cntfr; CNTFR alpha; CNTFRALPHA; CNTFR-alpha; MGC1774
Gene Symbols Cntfr
Host Species Goat
Immunogen Sf 21-derived recombinant human CNTF R alpha Gln23-Pro346 (predicted).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1271
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.