Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTNAP3 Polyclonal Antibody

CNTNAP3 Polyclonal Antibody

Supplier:  Thermo Scientific PA550641

Catalog No. PA550641


Only null left
Add to Cart

Description

Description

CNTNAP3 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CNTNAP3
Polyclonal
Unconjugated
CNTNAP3
CASPR3; Cell recognition molecule Caspr3; Cntnap3; CNTNAP3A; contactin associated protein-like 3; contactin-associated protein-like 3; KIAA1714; LRRGT00090; RGD1563615; RP11-138L21.1; RP11-290L7.1; similar to Contactin associated protein-like 3 precursor (Cell recognition molecule Caspr3)
Rabbit
Antigen Affinity Chromatography
RUO
79937
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9BZ76
CNTNAP3
Synthetic peptide corresponding to residues near the C terminal of human contactin associated protein-like 3.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.