Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNTNAP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561578
Description
Immunogen sequence: LHQVKINREE AVVMVEVNQS AKKQVILSSG TE Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
The specific function of this protein remains unknown. The protein encoded by this gene belongs to the NCP family of cell-recognition molecules. This family represents a distinct subgroup of the neurexins. NCP proteins mediate neuron-glial interactions in vertebrates and glial-glial contact in invertebrates. The protein encoded by this gene may play a role in cell recognition within the nervous system. Alternatively spliced transcript variants encoding different isoforms have been described but their biological nature has not been determined.
Specifications
CNTNAP3 | |
Polyclonal | |
Unconjugated | |
CNTNAP3 | |
CASPR3; Cell recognition molecule Caspr3; Cntnap3; CNTNAP3A; contactin associated protein-like 3; contactin-associated protein-like 3; KIAA1714; LRRGT00090; RGD1563615; RP11-138L21.1; RP11-290L7.1; similar to Contactin associated protein-like 3 precursor (Cell recognition molecule Caspr3) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79937 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZ76 | |
CNTNAP3 | |
Recombinant protein corresponding to Human CNTNAP3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction