Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTNAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103765 Supplier Invitrogen™ Supplier No. PA5103765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CNTNAP3.

The specific function of this protein remains unknown. The protein encoded by this gene belongs to the NCP family of cell-recognition molecules. This family represents a distinct subgroup of the neurexins. NCP proteins mediate neuron-glial interactions in vertebrates and glial-glial contact in invertebrates. The protein encoded by this gene may play a role in cell recognition within the nervous system. Alternatively spliced transcript variants encoding different isoforms have been described but their biological nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTNAP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CNTNAP3
Gene Accession No. Q9BZ76
Gene Alias CASPR3; Cell recognition molecule Caspr3; Cntnap3; CNTNAP3A; contactin associated protein-like 3; contactin-associated protein-like 3; KIAA1714; LRRGT00090; RGD1563615; RP11-138L21.1; RP11-290L7.1; similar to Contactin associated protein-like 3 precursor (Cell recognition molecule Caspr3)
Gene Symbols CNTNAP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CNTNAP3(Accession Q9BZ76), corresponding to amino acid residues L942-D992.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79937
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.