Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTNAP3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144361
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144361 Supplier Invitrogen™ Supplier No. PA5144361
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNTNAP3B Polyclonal Antibody for ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTNAP3B
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 4.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CNTNAP3B
Gene Accession No. Q96NU0
Gene Alias CASPR3B; Cell recognition molecule Caspr3b; CNTNAP3B; contactin associated protein-like 3B; contactin-associated protein-like 3B
Gene Symbols CNTNAP3B
Host Species Rabbit
Immunogen Recombinant Human Contactin-associated protein-like 3B protein (26-256AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728577
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.