Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTNAP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP186619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP186619 0.1 mL
NB431818 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP186619 Supplier Novus Biologicals Supplier No. NBP186619
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNTNAP4 Polyclonal specifically detects CNTNAP4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CNTNAP4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CASPR4KIAA1763cell recognition protein CASPR4, Cell recognition molecule Caspr4, contactin associated protein-like 4, contactin-associated protein-like 4
Gene Symbols CNTNAP4
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:CNMTETAWTIIQHNGSDLTRVRNTNPENPYAGFFEYVASMEQLQATINRAEHCEQEFTYYCKKSRLVN
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85445
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.