Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTNAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556881 Supplier Invitrogen™ Supplier No. PA556881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CNMTETAWTI IQHNGSDLTR VRNTNPENPY AGFFEYVASM EQLQATINRA EHCEQEFTYY CKKSRLVN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 72%.

Presynaptic protein involved in both dopaminergic synaptic transmission and GABAergic system, thereby participating in the structural maturation of inhibitory interneuron synapses. Involved in the dopaminergic synaptic transmission by attenuating dopamine release through a presynaptic mechanism. Also participates in the GABAergic system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTNAP4
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNTNAP4
Gene Accession No. Q9C0A0
Gene Alias Caspr4; cell recognition molecule Caspr4; cell recognition protein CASPR4; Cntnap4; contactin associated protein 4; contactin associated protein like 4; contactin associated protein-like 4; contactin-associated protein-like 4; E130114F09Rik; KIAA1763
Gene Symbols CNTNAP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNTNAP4. Recombinant protein control fragment (Product #RP-95657).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85445
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.