Learn More
Invitrogen™ CNTNAP4 Polyclonal Antibody

Description
Immunogen sequence: TESSCMAQPG TDATSRERTH SFADHSGTID DREPLANAIK SD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Specifications
Specifications
| Antigen | CNTNAP4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CNTNAP4 |
| Gene Accession No. | Q9C0A0 |
| Gene Alias | Caspr4; cell recognition molecule Caspr4; cell recognition protein CASPR4; Cntnap4; contactin associated protein 4; contactin associated protein like 4; contactin associated protein-like 4; contactin-associated protein-like 4; E130114F09Rik; KIAA1763 |
| Gene Symbols | CNTNAP4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.