Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTNAP4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548837
Description
Presynaptic protein involved in both dopaminergic synaptic transmission and GABAergic system, thereby participating in the structural maturation of inhibitory interneuron synapses. Involved in the dopaminergic synaptic transmission by attenuating dopamine release through a presynaptic mechanism. Also participates in the GABAergic system.
Specifications
| CNTNAP4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q9C0A0 | |
| CNTNAP4 | |
| KLH conjugated synthetic peptide between 1204-1231 amino acids from the C-terminal region of human CNTNAP4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CNTNAP4 | |
| Contactin-associated protein-like 4, Cell recognition molecule Caspr4, CNTNAP4, CASPR4, KIAA1763 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85445 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction