Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTNAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567043 Supplier Invitrogen™ Supplier No. PA567043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSVSINARRNR ITLTLDDEAA PPAPDSTWVQ IYSGNSYYFG GCPDNLTDSQ CLNPIKAFQG CM Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.

May play a role in the correct development and proper functioning of the peripheral and central nervous system and be involved in cell adhesion and intercellular communication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTNAP5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNTNAP5
Gene Accession No. Q8WYK1
Gene Alias CASPR5; cell recognition molecule Caspr5; CNTNAP5; contactin associated protein like 5; contactin associated protein-like 5; contactin-associated protein-like 5
Gene Symbols CNTNAP5
Host Species Rabbit
Immunogen Recombinant Human CNTNAP5. Recombinant protein control fragment (Product #RP-107687).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.