Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNTRL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562437
Description
Immunogen sequence: KTGVGTGANS QVLEIEKLNE TMERQRTEIA RLQNVLDLTG SDNKGGFENV LEEIAELRRE VSYQNDYISS MADPFKRRGY WYFMPPPPSS KVSSHSSQAT KDSGVG Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
This gene encodes a centrosomal protein required for the centrosome to function as a microtubule organizing center. The gene product is also associated with centrosome maturation. One version of stem cell myeloproliferative disorder is the result of a reciprocal translocation between chromosomes 8 and 9, with the breakpoint associated with fibroblast growth factor receptor 1 and centrosomal protein 1.
Specifications
CNTRL | |
Polyclonal | |
Unconjugated | |
CNTRL | |
110 kDa centrosomal protein; 6720467O09Rik; AA968343; antigen identified by monoclonal antibody 2A8; b2b1468Clo; bA165P4.1; bA165P4.1 (ortholog of mouse Ma2a8); bA165P4.2 (centrosomal protein 1); centriole associated protein; centriolin; Centrosomal protein 1; centrosomal protein 110; centrosomal protein 110kDa; centrosomal protein of 110 kDa; Cep1; CEP110; CNTRL; FAN; IB3/5; Ma2a8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z7A1 | |
CNTRL | |
Recombinant protein corresponding to Human CNTRL. Recombinant protein control fragment (Product #RP-92917). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction