Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554219 Supplier Invitrogen™ Supplier No. PA554219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAILERNENL EGELESLKEN LPFTMNEGPF EEKLNFSQVH IMDEHWRGEA LREKLRHRED RLKAQLRHCM SKQAEVLIKG KRQTEGTLHS LRRQVDALGE LVTST Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

This gene encodes a centrosomal protein required for the centrosome to function as a microtubule organizing center. The gene product is also associated with centrosome maturation. One version of stem cell myeloproliferative disorder is the result of a reciprocal translocation between chromosomes 8 and 9, with the breakpoint associated with fibroblast growth factor receptor 1 and centrosomal protein 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTRL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNTRL
Gene Accession No. Q7Z7A1
Gene Alias 110 kDa centrosomal protein; 6720467O09Rik; AA968343; antigen identified by monoclonal antibody 2A8; b2b1468Clo; bA165P4.1; bA165P4.1 (ortholog of mouse Ma2a8); bA165P4.2 (centrosomal protein 1); centriole associated protein; centriolin; Centrosomal protein 1; centrosomal protein 110; centrosomal protein 110kDa; centrosomal protein of 110 kDa; Cep1; CEP110; CNTRL; FAN; IB3/5; Ma2a8
Gene Symbols CNTRL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNTRL. Recombinant protein control fragment (Product #RP-92918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11064
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.