Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTROB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | CNTROB |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CNTROB Polyclonal specifically detects CNTROB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CNTROB | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
centrobin, centrosomal BRCA2 interacting protein, centrosomal BRCA2 interacting protein, Centrosomal BRCA2-interacting protein, LIP8centrobin, LYST-interacting protein 8, LYST-interacting protein LIP8, PP1221 | |
CNTROB | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
116840 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VTSQLYASLRLSRQAEATARAQLYLPSTSPPHEGLDGFAQELSRSLSVGLEKNLKKKDGSKHIFEMESVRGQLQTMLQTSRDTAYRDPLIPGAGSERREEDSFDSDSTATLLNTRPLQDLSPSSS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title