Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNTROB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554761
Description
Immunogen sequence: SQHSFQPLEP KPDLTSSTAG AFSALGAFHP DHRAERPFPE EDPGPDGEGL LKQGLPPAQL EGLKNFLHQL LETVPQNNEN Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 75%.
CNTROB is a centrosomal protein that interacts with BRCA2 and is required for centriole duplication and cytokinesis (Zou et al., 2005 ).
Specifications
CNTROB | |
Polyclonal | |
Unconjugated | |
CNTROB | |
9830165K03Rik; centrobin; centrobin, centriole duplication and spindle assembly protein; centrobin, centrosomal BRCA2 interacting protein; centrosomal BRCA2-interacting protein; CNTROB; LIP8; LYST-interacting protein 8; LYST-interacting protein LIP8; PP1221; RGD1307488 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116840 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N137 | |
CNTROB | |
Recombinant protein corresponding to Human CNTROB. Recombinant protein control fragment (Product #RP-93415). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction