Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTROB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554765 Supplier Invitrogen™ Supplier No. PA554765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHKTKVPLAM ASSLFRVPEP PSSHSQGSGP SSGSPERGGD GLTFPRQLME VSQLLRLYQA RGWGALPAED LLLYLKRLEH SGRTDGRGDN VPRRNTDSRL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 84%.

CNTROB is a centrosomal protein that interacts with BRCA2 and is required for centriole duplication and cytokinesis (Zou et al., 2005 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTROB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNTROB
Gene Accession No. Q8N137
Gene Alias 9830165K03Rik; centrobin; centrobin, centriole duplication and spindle assembly protein; centrobin, centrosomal BRCA2 interacting protein; centrosomal BRCA2-interacting protein; CNTROB; LIP8; LYST-interacting protein 8; LYST-interacting protein LIP8; PP1221; RGD1307488
Gene Symbols CNTROB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNTROB. Recombinant protein control fragment (Product #RP-93416).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116840
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.