Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C15orf40 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548417
Description
Predicted to react with bovine, mouse and rat based on sequence homology.

Specifications
C15orf40 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8WUR7 | |
C15ORF40 | |
KLH conjugated synthetic peptide between 72-100 amino acids from the C-terminal region of human CO040. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
C15orf40 | |
UPF0235 protein C15orf40, C15orf40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
123207 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction