Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heratherm CO2 Chamber

Catalog No. 01-672-444PM
Click to view available options
Door Style:
Glass Door
Solid Outer, Glass Inner

Catalog No. 01-672-444PM

Supplier: Thermo Scientific™ ECS750PM

Precisely control the essential parameters of your distinct cell growth or CO2 testing conditions - active Humidity controls the incubation environment with accuracy.

  • Uniform Temperature / CO2 / Relative Humidity throughout the chamber
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • CO control system ensures stable CO2 environment, range of 0 to 20 Vol-%.
  • Environmentally friendly water-efficient humidification system - with water tank or optional fixed water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option.
  • Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Thermo Cloud via OPC UA
Capacity (Metric) 750 L
Chamber Material Stainless Steel (304/1.4301)
Material Powder-coated Galvanized Steel
No. of Shelves 3
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Type Growth Chamber
Display Touchscreen
Door Style Solid Outer, Glass Inner
For Use With (Application) Cell growth, testing
Humidity Range 10 to 90% RH
Humidity Resolution 0.1% RH
Humidity Source Removable water tank
No. of Casters 4
Ports 2 Access Ports
Programmability Temperature, Relative Humidity, CO2 Ramping
Shelf Area (English) 4.9 ft.2
Shelf Area (Metric) 0.46 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor
Warranty 24 Months
Water Consumption <0.8 L/day
Water Resistivity 0.05 to 1 MOhm
Dimensions (L x W x H) 35.24 x 42.52 x 72.24 in. (89.5 x 108 x 183.5 cm)
Dimensions (LxWxH) Interior 24.8 x 33.07 x 57.09 in. (63 x 84 x 145 cm)
Width (English) Shelf 31.3 in.
Width (Metric) Shelf 79.5 cm
Shipping Weight (English) 859 lb.
Shipping Weight (Metric) 390 kg
Electrical Requirements 100/240 V, 50/60 Hz, 1400 W
Voltage 100/115/220/230 V
Wattage 1400 W
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.